Learn More
CD45.2 Mouse anti-Mouse, PE-Cy7, Clone: 104, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 560696
Description
The 104 clone has been reported to react with CD45 (Leukocyte Common Antigen) on all leukocytes of most mouse strains (eg, A, AKR, BALB/c, CBA/Ca, CBA/J, C3H/He, C57BL, C57BR, C57L, C58, DBA/1, DBA/2, NZB, SWR, 129). This alloantigen was originally named Ly-5.1, and this was the designation at the time that the antibody was characterized. The designation was later changed from Ly-5.1 to Ly-5.2 to conform with the convention that the .2 alloantigen designations be assigned to the C57BL/6 strain. mAb 104 has been reported not to react with leukocytes of the mouse strains expressing the CD45.1 alloantigen (eg, RIII, SJL/J, STS/A, and DA). CD45 is a member of the Protein Tyrosine Phosphatase (PTP) family: its intracellular (COOH-terminal) region contains two PTP catalytic domains, and the extracellular region is highly variable due to alternative splicing of exons 4, 5, and 6 (designated A, B, and C, respectively), plus differing levels of glycosylation. The CD45 isoforms detected in the mouse are cell type-, maturation-, and activation state-specific. The CD45 isoforms play complex roles in T-cell and B-cell antigen receptor signal transduction. The 104 antibody has been reported to inhibit some responses of B cells, from mice expressing the CD45.2 alloantigen, to certain antigens and LPS. In addition, reduction of serum IgG levels and amelioration of autoimmune renal pathology were reported in mAb 104-treated systemic lupus erythematosus-prone mice.
Specifications
| CD45.2 | |
| Monoclonal | |
| 0.2mg/mL | |
| Ly-5.2; T200; LCA; Leukocyte common antigen; Ptprc | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| 104 | |
| PE-Cyanine7 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| B10.S mouse thymocytes and splenocytes | |
| 50 μg | |
| Immunology | |
| Mouse | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.