missing translation for 'onlineSavingsMsg'
Learn More

CD8 Mouse anti-Human, APC-H7, Clone: SK1, BD

Mouse Monoclonal Antibody

181.00€ - 454.00€

Specifications

Antigen CD8
Clone SK1
Applications Flow Cytometry
Classification Monoclonal
Conjugate APC-H7
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15830069
View Documents
BD Biosciences
560273
25 Tests
181.00€
Each
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12880500
View Documents
BD Biosciences
561423
50 Tests
287.00€
Each
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15879988
View Documents
BD Biosciences
560179
100 Tests
454.00€
Each
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD8 recognizes the 32kDa a-subunit of a disulfide-linked bimolecular complex. The majority of peripheral blood CD8+ T lymphocytes express an a/b heterodimer (Mr 32, 30kDa), while CD8+CD16+ natural killer (NK) lymphocytes and CD8+ T-cell receptor (TCR)-γ/δ+ lymphocytes express a/a homodimer (Mr 30kDa). CD8+TCR-α/β+ lymphocytes can express either an α/α homodimer or α/β heterodimer. The CD8 antigenic determinant binds to class I major histocompatibility (MHC) molecules resulting in increased adhesion between the CD8+ T lymphocytes and target cells. Binding of the CD8 antigen is coupled to a protein tyrosine kinase p56lck. The CD8:p56lck complex can play a role in T-lymphocyte activation through mediation of the interactions between the CD8 antigen and the CD3/TCR complex.

Host Species: Mouse
Clone: SK1
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Human Peripheral Blood T Cells

Flow Cytometry

Specifications

Specifications

CD8
Flow Cytometry
APC-H7
RUO
Baboon, Cynomolgus, Human, Rhesus Monkey
Human Peripheral Blood T Cells
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
SK1
Monoclonal
Mouse
Immunology
Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide.
IgG1 κ
Affinity Purified
CD8α; CD8A; CD8 alpha; Leu2a; MAL; T8; p32
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.