missing translation for 'onlineSavingsMsg'
Learn More

CD82 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA513228

Additional Details : Weight : 0.01000kg

Product Code. 10668370

  • 506.00€ / 100µg
Estimated Shipment: 08-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CD82 Polyclonal antibody specifically detects CD82 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.
Specifications

Specifications

CD82
Polyclonal
Unconjugated
CD82
4F9; AA682076; AL023070; C33; C33 antigen; Cd82; CD82 antigen; CD82 antigen (R2 leukocyte antigen antigen detected by monoclonal and antibody IA4)); CD82 antigen (R2 leukocyte antigen, antigen detected by monoclonal and antibody IA4)); CD82 molecule; GR15; IA4; Inducible membrane protein R2; KAI1; Kangai 1 (suppression of tumerigenicity 6 prostate) CD82 antigen; kangai 1 (suppression of tumorigenicity 6), prostate; kangai 1 (suppression of tumorigenicity 6, prostate); Kangai 1 (suppression of tumorigenicity 6, prostate; CD82 antigen (R2 leukocyte antigen, antigen detected by monoclonal and antibody IA4)); metastasis suppressor Kangai-1; Metastasis suppressor Kangai-1 homolog; R2; SAR2; ST6; Suppressor of tumorigenicity 6 protein; Tetraspanin-27; Tspan27; tspan-27
Rabbit
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
12521, 3732
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
1.7 mg/mL
PBS with 0.09% sodium azide
P27701, P40237
CD82
KLH conjugated synthetic peptide between 239-267 amino acids from the C-terminal region of human CD82 (ST6)
400 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions