Learn More
CD95 Mouse anti-Human, Brilliant Violet 650, Clone: DX2, BD Optibuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 740589
Description
The DX2 monoclonal antibody specifically binds to the human Fas antigen (also called APO-1). This 45 kDa transmembrane cell surface molecule was designated as CD95 at the Fifth HLDA Workshop. Fas is a member of the TNF-receptor superfamily and is also known as Tumor necrosis factor receptor superfamily member 6 (TNFRSF6). It is expressed on a variety of normal and neoplastic cells including activated T and B lymphocytes and some undifferentiated thymocytes. The Fas/CD95 antigen is a polypeptide that plays a role in the programmed sequence of events leading to cell death, termed apoptosis. The DX2 clone specifically reacts with murine L cells, murine L1210 leukemia cells and murine P815 mastocytoma cells transfected with human Fas cDNA but not with untransfected parental cell lines. Crosslinking CD95 with DX2 antibody delivers an apoptotic signal indicating that DX2 recognizes a functional epitope of the CD95 antigen.
The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 650-nm. BD Horizon BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination.
Specifications
| CD95 | |
| Monoclonal | |
| 0.2mg/mL | |
| APO-1; FAS; TNFRSF6; APT1; ALPS1A; FAS1; FASTM; FASLG receptor | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| DX2 | |
| Brilliant Violet 650 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Human CD95-transfected L Cells | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only