Learn More
CLIP Mouse anti-Human, FITC, Clone: CERCLIP, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 555981
Additional Details : Weight : 0.02000kg
Description
Reacts with the class II associated invariant chain peptides (CLIP). CLIP is the remaining segment of class II invariant chain (Ii) after proteolytic degradation which stays associated with HLA-DR and represents a final intermediate in the removal of invariant chain from class II molecules during antigen processing. The removal of CLIP and subsequent loading of the antigenic peptide is facilitated by the non-classical class II molecule HLA-DM. CLIP can be detected, in association with HLA-DR, on the surface of T2DR3, a mutant cell line lacking HLA-DM.
Host Species: Mouse
Clone: CerCLIP
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry
Specifications
CLIP | |
Monoclonal | |
FITC | |
Mouse | |
100 Tests | |
Immunology | |
Human | |
IgG1 κ |
Flow Cytometry | |
CERCLIP | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.