missing translation for 'onlineSavingsMsg'
Learn More

GBF1 (Golgi-specific Brefeldin A (BFA)-resistance Factor 1) Mouse, Unlabeled, Clone: 25, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 612116

Product Code. 15846099

  • ONLINE EXCLUSIVE PRICE
    187.00€
    186.18€ / 1 set
    Save 0.82€
    0.44% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The Golgi apparatus is a complex and dynamic organelle that functions in protein sorting and modification. Numerous structural and regulatory proteins are involved in the budding, docking, and fusion of Golgi-directed vesicles. Golgi-specific Brefeldin A (BFA)-resistance Factor (GBF1) is a guanine nucleotide exchange factor that contains a centrally located Sec7 domain, and a proline-rich region at the C-terminus. The Sec7 domain is commonly found in a variety of secretory proteins, and is required for activation of ARFs. GBF1 mRNA is ubiquitously expressed, and GBF1 protein localizes to the Golgi, as well as to perinuclear structures that contain COPI. BFA is a fungal heterocyclic lactone that disrupts membrane recruitment of ARFs. This action blocks protein secretion, and changes the morphology of various organelles, such as the Golgi. Overexpression of GBF1 suppresses BFA-induced changes in Golgi morphology, ARF activation, and coat protein recruitment. GBF1 interacts preferentially with ARF5 in the presence of Mg2+;. Thus, GBF1 is a Golgi-specific guanine nucleotide exchange factor that activates ARFs through a mechanism that interferes with BFA action.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

GBF1 (Golgi-specific Brefeldin A (BFA)-resistance Factor 1)
Monoclonal
250μg/mL
Golgi-specific Brefeldin A (BFA)-resistance Factor 1
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Western Blot
25
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Human GBF1 aa. 1266-1379
50 μg
Cell Biology
Human, Mouse, Rat
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.