missing translation for 'onlineSavingsMsg'
Learn More

anti-H2AX; Clone: N1-431; BD

Brand:  BD Biosciences 562377

Product Code. 15801869

  • ONLINE EXCLUSIVE PRICE
    276.00€
    275.35€ / Each
    Save 0.65€
    0.24% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Host Species: Mouse
Clone: N1-431
Isotype: IgG1, κ
Species Reactivity: Hu, Ms
Immunogen: Phosphorylated Human H2AX Peptide

Flow Cytometry (Intracellular Staining)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

H2AX
N1-431
Mouse
Affinity Purified
RUO
Human, Mouse
Monoclonal
PE
Phosphorylated Human H2AX Peptide
50 Tests
Primary
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.