Description
- Produced from a monoclonal cell line cultivated in mice (Host) and provided as clarified ascites fluid
- Provides sufficient antibody (monoclonal) for 25 Western blots
- IgG2 subclass of monoclonal antibody directed against six histidine residues, which are commonly used as an affinity tag for recombinant fusion proteins
- Offers the flexibility of detection with a secondary antibody conjugated with either horseradish peroxidase or alkaline phosphatase
- Provides high sensitivity with low background
- Volume 170μL
ELISA, Western Blotting
Specifications
Specifications
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Quantity | 170 μL |
| Primary or Secondary | Primary |
| Form | Ascites |
| Isotype | IgG2 |