missing translation for 'onlineSavingsMsg'
Learn More

Anti-Human CD226 (11A8), Proteintech

Mouse Monoclonal Antibody

60.00€ - 161.00€

Specifications

Antigen CD226
Clone 11A8
Concentration 0.5 mg/mL
Applications Flow Cytometry, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30234070
View Documents
Proteintech
65247-1-IG-500UG
500 μg
ONLINE EXCLUSIVE PRICE
173.00€
161.00€ / 500µg
Save 12.00€
6% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30234695
View Documents
Proteintech
65247-1-IG-100UG
100 μg
ONLINE EXCLUSIVE PRICE
64.00€
60.00€ / 100µg
Save 4.00€
6% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The CD226 antibody from Proteintech is a mouse monoclonal antibody generated with human nk cells of human CD226. This antibody recognizes human antigen. The CD226 antibody has been validated for the following applications: FC, ELISA analysis.
Specifications

Specifications

CD226
0.5 mg/mL
Monoclonal
Liquid
RUO
10666
Human NK Cells
Primary
Store at 2-8°C. Stable for one year after shipment.
11A8
Flow Cytometry, ELISA
Unconjugated
Mouse
Human
DNAM-1; DNAM1; PTA1; TLiSA1
IgG1 κ
Affinity purification
Flow Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.