missing translation for 'onlineSavingsMsg'
Learn More

Anti-Human CD33 (WM53), Proteintech

Mouse Monoclonal Antibody

53.00€ - 158.00€

Specifications

Antigen CD33
Clone WM53
Concentration 0.5 mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30236359
View Documents
Proteintech
65272-1-IG-100UG
100 μg
53.00€
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30236028
View Documents
Proteintech
65272-1-IG-500UG
500 μg
ONLINE EXCLUSIVE PRICE
163.00€
158.00€ / 500µg
Save 5.00€
3% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The CD33 antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human CD33. This antibody recognizes human antigen. The CD33 antibody has been validated for the following applications: FC analysis.
Specifications

Specifications

CD33
0.5 mg/mL
Monoclonal
Liquid
RUO
945
IgG1 κ
Affinity purification
Flow Antibody
WM53
Flow Cytometry
Unconjugated
Mouse
Human
FLJ00391; SIGLEC-3; SIGLEC3; p67
Primary
Store at 2-8°C. Stable for one year after shipment.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.