missing translation for 'onlineSavingsMsg'
Learn More

Anti-Human CD85j / LILRB1 (GHI/75), Proteintech

Mouse Monoclonal Antibody

63.00€ - 195.00€

Specifications

Antigen CD85j / LILRB1
Clone GHI/75
Concentration 0.5 mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30233963
View Documents
Proteintech
65238-1-IG-100UG
100 μg
ONLINE EXCLUSIVE PRICE
68.00€
63.00€ / 100µg
Save 5.00€
7% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30237067
View Documents
Proteintech
65238-1-IG-500UG
500 μg
ONLINE EXCLUSIVE PRICE
209.00€
195.00€ / 500µg
Save 14.00€
6% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The CD85j / LILRB1 antibody from Proteintech is a mouse monoclonal antibody generated with human hairy cell leukemia spleen cells of human CD85j / LILRB1. This antibody recognizes human antigen. The CD85j / LILRB1 antibody has been validated for the following applications: FC analysis.
Specifications

Specifications

CD85j / LILRB1
0.5 mg/mL
Monoclonal
Liquid
RUO
10859
Human Hairy Cell Leukemia Spleen Cells
Primary
Store at 2-8°C. Stable for one year after shipment.
GHI/75
Flow Cytometry
Unconjugated
Mouse
Human
CD85; CD85J; FLJ37515; ILT2; LIR-1; LIR1; MIR-7; MIR7
IgG2b κ
Affinity purification
Flow Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.