missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ID1 Monoclonal Antibody (3F3), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA519170
Additional Details : Weight : 0.00010kg
Description
Peptide Sequence: GGAGARLPAL LDEQQVNVLL YDMNGCYSRL KELVPTLPQN RKVSKVEILQ HVIDYIRDLQ LELNSESEVG TPGGRGLPVR APLSTLNGEI SALTAEAACV PADDRILCR*
ID1 Monoclonal antibody specifically detects ID1 in Human samples. It is validated for ELISA, Immunocytochemistry, Western Blot.Specifications
ID1 | |
Monoclonal | |
1.0 mg/mL | |
PBS with no preservative; pH 7.4 | |
P41134 | |
ID1 | |
ID1 (NP_002156, 47 a.a. ∽ 155 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Immunocytochemistry, Western Blot | |
3F3 | |
Unconjugated | |
ID1 | |
AI323524; bHLHb24; class B basic helix-loop-helix protein 24; D2Wsu140e; dJ857M17.1.2; dJ857M17.1.2 (inhibitor of DNA binding 1, dominant negative helix-loop-helix protein); DNA-binding protein inhibitor ID-1; ID; ID1; Id-1; ID125A; Idb1; inhibitor of differentiation 1; Inhibitor of DNA binding 1; Inhibitor of DNA binding 1 helix-loop-helix protein (splice varaiation); inhibitor of DNA binding 1, dominant negative helix-loop-helix protein; Inhibitor of DNA binding 1, helix-loop-helix protein (splice variation); inhibitor of DNA binding 1, HLH protein | |
Mouse | |
Protein A | |
RUO | |
3397 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |