missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG (Multiple Adsorption) Donkey anti-Rabbit, PE, Polyclonal, BD
Donkey Polyclonal Antibody
Brand: BD Biosciences 558416
This item is not returnable.
View return policy
Description
The polyclonal F(ab')2 antibody reacts with heavy chains of rabbit IgG and light chains of most rabbit immunoglobulins, and not with non-immunoglobulin proteins of rabbit serum. It has minimal cross-reactivity with human, mouse, rat, bovine, caprine, equine, ovine, chicken, guinea pig, and Syrian hamster serum proteins.
Control, Intracellular Staining
Specifications
IgG (Monkey adsorbed) | |
Polyclonal | |
PE | |
Donkey | |
50 Tests | |
Immunology | |
Rabbit | |
Liquid |
Flow Cytometry | |
POLYCLONAL | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Rabbit IgG Protein | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG |