missing translation for 'onlineSavingsMsg'
Learn More

anti-IgM κ, Mouse, FITC, Clone: G155-228, Isotype Control, BD

Product Code. 15826658
Change view
Click to view available options
Quantity:
0.25 mg
50 Tests
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15826658 50 Tests Each
15847238 0.25 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15826658 Supplier BD Biosciences Supplier No. 551448

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The G155-228 antibody is specific for the hapten trinitrophenol (TNP), a hapten not expressed on non-human primate cells or non-human primate cell lines. In the absence of specific binding, this immunoglobulin may bind non-specifically to Fc receptors.

Host Species: Mouse
Clone: G155-228
Isotype: IgM, κ
Species Reactivity: Non-human primate
Immunogen: TNP-Keyhole Limpet Hemocyanin

Flow Cytometry, Immunofluorescence, Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Applications Flow Cytometry, Immunofluorescence, Western Blot
Clone G155-228
Conjugate FITC
Host Species Mouse
Immunogen TNP-keyhole limpet hemocyanin
Purification Method Affinity Purified
Quantity 50 Tests
Target Species Non-human Primate
Content And Storage Undiluted at 4°C. Protect from exposure to light. Do not freeze.
Isotype IgM κ
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.