missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Light Chain, l Mouse anti-Human, PE, Clone: JDC-12, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 562054
This item is not returnable.
View return policy
Description
The JDC-12 monoclonal antibody specifically binds to human immunoglobulin light chain, lambda (μ). It does not not bind to immunoglobulin κ light chains or heavy chains.
Specifications
| Light Chain, l | |
| Monoclonal | |
| PE | |
| Mouse | |
| 25 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| JDC-12 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction