missing translation for 'onlineSavingsMsg'
Learn More

Ly-6G and Ly-6C Rat anti-Mouse, Biotin, Clone: RB6-8C5, BD

Rat Monoclonal Antibody

Brand:  BD Biosciences 553124

 View more versions of this product

Product Code. 15897008

  • 229.00€ / Each

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The RB6-8C5 antibody reacts with a common epitope on Ly-6G and Ly-6C, previously known as the myeloid differentiation antigen Gr-1. In the bone marrow, the level of antigen expression is directly correlated with granulocyte differentiation and maturation.

Host Species: Rat
Clone: RB6-8C5
Isotype: IgG2b, κ
Species Reactivity: Mouse

Cell Separation, Flow Cytometry

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Ly-6G and Ly-6C
Monoclonal
0.5mg/mL
Ly6c, Lymphocyte antigen 6C2; Lymphocyte antigen 6G, Ly6g, Gr-1
Rat
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
RB6-8C5
Biotin
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Not Reported
0.1 mg
Immunology
Mouse
IgG2b κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.