missing translation for 'onlineSavingsMsg'
Learn More

NCK Mouse, Unlabeled, Clone: 108, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 610099

 View more versions of this product

Product Code. 15835259

  • ONLINE EXCLUSIVE PRICE
    251.00€
    249.85€ / 1 set
    Save 1.15€
    0.46% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

NCK is a 47kDa protein that belongs to a class of signaling molecules called adaptors. These proteins exert their effects by coupling activated receptors to downstream effectors. NCK is comprised almost entirely of one SH2 and three SH3 domains and it possesses no intrinsic catalytic activity. Investigations of its interactions with cellular proteins have found constitutive association of NCK with various Ser/THr kinases, such as p21-associated kinase (PAK) and casein kinase 1. Other proteins that directly interact with NCK are: Sos, Cbl, Wiskott-Aldrich syndrome protein (WASP), IRS1, and multiple tyrosine kinase receptors. The presence of NCK is necessary for the mitogenic response elicited by PDGF in some cells. In addition, the Drosophila homologue of NCK is involved in regulating cytoskeletal organization.

Host Species: Mouse
Clone: 108
Isotype: IgG2b
Species Reactivity: Human
Immunogen: Human NCK aa. 279-377
Formula Weight [Chemical]: 47kDa

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

NCK
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Human NCK aa. 279-377
50 μg
Cell Biology
Bovine, Canine, Chicken, Frog, Human, Mouse, Rat
IgG2b
Western Blot
108
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.