missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFV2, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human NDUFV2 protein.
Brand: Abnova H00004729-B01P.50ug
This item is not returnable.
View return policy
Description
Sequence: MFFSAALRARAAGLTAHWGRHVRNLHKTAMQNGAGGALFVHRDTPENNPDTPFDFTPENYKRIEAIVKNYPEGHKAAAVLPVLDLAQRQNGWLPISAMNKVAEVLQVPPMRVYEVATFYTMYNRKPVGKYHIQVCTTTPCMLRNSDSILEAIQKKLGIKVGETTPDKLFTLIEVECLGACVNAPMVQINDNYYEDLTAKDIEEIIDELKAGKIPKPGPRSGRFSCEPAGGLTSLTEPPKGPGFGVQAGLSpecifications
| NDUFV2 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human NDUFV2 protein. | |
| NDUFV2 | |
| NDUFV2 | |
| NDUFV2 (NP_066552.1, 1 a.a. ∼ 249 a.a) full-length human protein. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_021074 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4729 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction