missing translation for 'onlineSavingsMsg'
Learn More

NMDAR2B Mouse, Unlabeled, Clone: 13, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 610416

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 15384742

  • 330.00€ / Each
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The majority of synapses in the central nervous system utilize glutamate as a neurotransmitter to produce rapid neuronal excitation. Glutamate has a diverse array of receptors that can be categorized into two groups: ionotropic and metabotropic. The ionotropic receptors are subdivided into two distint types: 1) receptors for N-methyl D-aspartate (NMDAR) and 2) non-NMDA receptors for AMPA and kainate. Three types of NMDAR2 have been identified: NR2A, NR2B, and NR2C. NR2A and NR2B contain a C-terminal extension (>600 amino acids) that has small scattered regions of conserved sequence. The three NR2 mRNAs show overlapping, differential expression patterns in the rat brain. NR2B has been reported to be expressed in the forebrain, thalamic nuclei, amygdaloid nuclei, caudateputamen, and in restricted regions of the olfactory bulb.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

NMDAR2B
Monoclonal
250μg/mL
N-Methyl-D-Aspartate Receptor 2B
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Western Blot
13
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
50 μg
Cell Biology
Human, Mouse, Rat
IgG2b
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

For Research Use Only.