missing translation for 'onlineSavingsMsg'
Learn More

Paxillin Mouse, Unlabeled, Clone: 177, BD

Mouse Monoclonal Antibody

175.91€ - 417.30€

Specifications

Antigen Paxillin
Clone 177
Concentration 250μg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15805529
View Documents
BD Biosciences
610568
50 μg
ONLINE EXCLUSIVE PRICE
176.00€
175.91€ / 1 set
Save 0.09€
0.05% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15815529
View Documents
BD Biosciences
610569
150 μg
ONLINE EXCLUSIVE PRICE
419.00€
417.30€ / 1 set
Save 1.70€
0.41% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Paxillin, a focal adhesion protein, is a substrate for several tyrosine kinases such as src, FAK, and p120BRC/ABL. The tyrosine phosphorylation of paxillin is affected by conditions that change cell-cell adhesion. This is consisent with the possibility that paxillin is involved in the regulation of cell morphology. Additionally, because of its SH3-binding domain, paxillin associates tightly with FAK and Crk in an extracellular matrix-independent manner. Although paxillin was initially detected in fibroblasts, its phosphorylation may be important during neurite extension during differentiation.

Specifications

Specifications

Paxillin
250μg/mL
Monoclonal
Mouse
Cell Biology
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
IgG1
Affinity Purified
177
Western Blot
Unconjugated
RUO
Bovine, Canine, Chicken, Human, Mouse, Rat
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Primary
Store undiluted at -20°C.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.