missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PI3-Kinase Mouse, Unlabeled, Clone: 4, BD
Mouse Monoclonal Antibody
450.58€
Specifications
| Antigen | PI3-Kinase |
|---|---|
| Clone | 4 |
| Concentration | 250μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
PI3-kinase phosphorylates the D-3 position of the inositol ring of phosphatidylinositol (PtdIns), PtdIns(4)P and PtdIns(4,5)P2 to produce the respective PI3-phosphorylated derivatives. PI3-kinase exists as a heterodimer of 85 kDa (p85) and 110 kDa (p110) subunits. The p85 subunit contains two SH2 domains and an SH3 domain. It associates with and serves as a substrate for activated growth factor receptor tyrosine kinases. p85 may serve as regulator of the catalytic subunit, p110, by acting as the link between PI3-kinase and the ligand-activated receptor. Two distinct forms of the p85 subunit have been described: 1) p85 α; , which binds tightly to the catalytic subunit, and 2) p85ß, a protein whose function is presently unknown. Both isoforms bind to activated receptors and serve as tyrosine kinase substrates.Specifications
| PI3-Kinase | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| IgG2a | |
| Affinity Purified |
| 4 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Canine, Chicken, Human, Mouse, Rat | |
| Human PI3-Kinase α subunit aa. 562 to 724 | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Name des Produkts