missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-Rat IgG2b, kappa Isotype Control, Clone: R35-38, BD
Description
Host Species: Rat
Clone: R35-38
Isotype: IgG2b, kappa
Species Reactivity: Hu, Ms
Isotype Control
Specifications
Specifications
| Antigen | Rat IgG2b, kappa |
| Classification | Monoclonal |
| Clone | R35-38 |
| Concentration | 0.2mg/mL |
| Conjugate | Brilliant Violet 786 |
| Host Species | Rat |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Regulatory Status | RUO |
| Primary or Secondary | Secondary |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?