Learn More
SMAD1 Recombinant Rabbit Monoclonal Antibody (12H1L14), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen 701168
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with equine, mouse, rabbit and rat based on sequence homology. Intact IgG appears on a non-reducing gel as ∽150 kDa band and upon reduction generating a ∽25 kDa light chain band and a ∽50 kDa heavy chain. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
SMAD1 Recombinant Monoclonal antibody specifically detects SMAD1 in Human, Mouse samples. It is validated for ChIP Assay, Western Blot.Specifications
SMAD1 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
AI528653; BSP1; BSP-1; dwarfin-A; Dwf-A; DWFC; hSMAD1; JV41; JV4-1; JV5-1; MAD (mothers against decapentaplegic, Drosophila) homolog 1; MAD homolog 1; MAD homolog1 (mothers against decapentaplegic, Drosophila); MAD, mothers against decapentaplegic homolog 1; Madh1; MADH5; MADR1; Mad-related protein 1; Mlp1; mMad1; mothers against decapentaplegic homolog 1; mothers against DPP homolog 1; mothers-against-DPP-related 1; Mothers-against-DPP-related-1; MusMLP; OTTHUMP00000220456; SMAD; Smad 1; SMAD family member 1; SMAD, mothers against DPP homolog 1; Smad1; SMAD5; TGF-beta signaling protein 1; transforming growth factor-beta signaling protein 1; Transforming growth factor-beta-signaling protein 1 | |
Peptide corresponding to amino acids 178–193 of human SMAD1 | |
100 μg | |
Primary | |
Human, Mouse | |
Liquid |
ChIP Assay, Western Blot | |
12H1L14 | |
Unconjugated | |
P70340, Q15797 | |
Rabbit | |
Protein A | |
RUO | |
17125, 4086 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |