Learn More
Sox2 Mouse, PE, Clone: O30-678, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 562195
Description
The monoclonal antibody O30-678 recognizes the Sox2 transcription factor. Sox2[SRY (sex determining region Y)-box 2] is a member of theSRY-related HMG-box (SOX) family of transcription factors. Sox2 is required for the maintenance of the undifferentiated state of pluripotent stem cells. Complexes of Sox2 with the homeobox transcription factors Oct3/4 and/or Nanog bind to the promoters of a network of genes that are involved in the maintenance of pluripotency and self renewal in stem cells. Sox2 is also a marker of neural stem cells during embryonic development and in the adult brain. The O30-678 antibody recognizes both human and mouse Sox2 proteins.
Specifications
| Sox2 | |
| Monoclonal | |
| PE | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human, Mouse | |
| IgG1 κ |
| Flow Cytometry | |
| O30-678 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human Sox2 Recombinant Protein | |
| 50 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.