Learn More
CD248 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA520437
Additional Details : Weight : 0.01000kg
Description
A suggested positive control is human colon tissue lysate. PA5-20437 can be used with blocking peptide PEP-0554. The PA5-20437 immunogen is located within amino acids 40-90 of TEM-1. Predicted molecular ∽ 81kD. In Western blot applications, this antibody has been observed to detect a band at: 90 kD (post-modifications: 27 O-linked glycosylations). Predicted species reactivity based on immunogen sequence: Mouse (100%); Rat: (93%).
CD248 Polyclonal antibody specifically detects CD248 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
CD248 | |
Polyclonal | |
Unconjugated | |
CD248 | |
2610111G01Rik; AI842296; CD164 sialomucin-like 1; CD164L1; Cd248; CD248 antigen, endosialin; CD248 molecule; CD248 molecule, endosialin; endosialin; MGC119478; MGC119479; TEM1; tumor endothelial marker 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57124 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q9HCU0 | |
CD248 | |
A 14 amino acid peptide near the amino terminus of the human TEM1. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |