Learn More
Tet Repressor Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA129677
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: Tetracyclin repressor (tetR). This antibody works in any species where the TetR system is being used to regulate gene ezpression in mammalian cells. Not suitable for immunofluorescence.
Tet Repressor Polyclonal antibody specifically detects Tet Repressor in Bacteria samples. It is validated for ELISA, Western Blot.Specifications
Tet Repressor | |
Polyclonal | |
Unconjugated | |
pSC101 tetR; TetR | |
Reacts with residues 31-49 [AQKLGVEQPTLYWHVKNKR] of the 22 kDa Tet repressor protein. Sequence is 100% conserved in all TetR classes. | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
1.36 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
Bacteria | |
Antibody | |
IgG |
Safety and Handling
- Tet Repressor Polyclonal Antibody
Signal Word
- Warning
Hazard Category
- Skin sensitiser Category 1A
Hazard Statement
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
- MIXTURE LIST-Contains : 2-methylisothiazol-3(2H)-one