missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TET3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA534431
Additional Details : Weight : 0.01000kg
Description
A suggested positive control is SK-N-SH cell lysate. PA5-34431 can be used with blocking peptide PEP-1473.
TET3 Polyclonal antibody specifically detects TET3 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.Specifications
TET3 | |
Polyclonal | |
Unconjugated | |
TET3 | |
B430006D22Rik; BC037432; D230004J03Rik; hCG_40738; KIAA0401; Methylcytosine dioxygenase TET3; MGC22014; probable methylcytosine dioxygenase TET3; putative methylcytosine dioxygenase; Tet methylcytosine deoxygenase 3 isoform; tet methylcytosine dioxygenase 3; tet oncogene family member 3; Tet3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
194388, 200424, 680576 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
O43151, Q8BG87 | |
TET3 | |
A 17 amino acid synthetic peptide near the carboxy terminus of human TET3 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |