missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAR1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA534263
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human (100%) Baboon, Monkey (94%) Gorilla, Gibbon (89%) Marmoset (83%).
Thrombin is a serine protease that is involved in platelet aggregation and blood coagulation. It is cleaved from its precursor, prothrombin, and converts fibrinogen to fibrin in the final step of the clotting cascade. Thrombin mediates its regulatory effects by activating cell surface G-protein coupled receptors which include PAR-1, PAR-2 and PAR3.Specifications
PAR1 | |
Polyclonal | |
Unconjugated | |
F2R | |
AI482343; C HTR; Cf2r; coagulation factor II (thrombin) receptor; coagulation factor II receptor; coagulation factor II thrombin receptor; F2R; HTR; PAR1; PAR-1; protease activated receptor 1; protease-activated receptor 1; proteinase-activated receptor 1; Thrombin receptor; ThrR; TR; TRGPC | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2149 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P25116 | |
F2R | |
Synthetic 18 amino acid peptide from N-terminal extracellular domain of human Thrombin Receptor. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |