missing translation for 'onlineSavingsMsg'
Learn More

Thy-1 cell surface antigen, Mouse, Polyclonal Antibody, Abnova™

Product Code. 16101382
Change view
Click to view available options
Quantity:
50 μL
Unit Size:
50µL
Product Code. Quantity unitSize
16101382 50 μL 50µL
1 options
This item is not returnable. View return policy

Product Code. 16101382

Brand: Abnova H00007070B01.50uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse polyclonal antibody raised against a full-length human THY1 protein.

Sequence: MNLAISIALLLTVLQVSRGQKVTSLTACLVDQSLRLDCRHENTSSSPIQYEFSLTRETKKHVLFGTVGVPEHTYRSRTNFTSKYNMKVLYLSAFTSKDEGTYTCALHHSGHSPPISSQNVTVLRDKLVKCEGISLLAQNTSWLLLLLLSLSLLQATDFMSL

Specifications

Antigen Thy-1 cell surface antigen
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human THY1 protein.
Formulation No additive
Gene THY1
Gene Accession No. BC005175
Gene Alias CD90/FLJ33325
Gene Symbols THY1
Host Species Mouse
Immunogen THY1 (AAH05175, 1 a.a. ∼ 161 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Stem Cell Biology
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 7070
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.