missing translation for 'onlineSavingsMsg'
Learn More

UNC5B Polyclonal Antibody, Invitrogen™

Goat Polyclonal Antibody

Brand:  Invitrogen PA537993

Additional Details : Weight : 0.00900kg

Product Code. 15574921

  • 465.00€ / 100µg
Estimated Shipment: 08-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This antibody is predicted to react with mouse, rat, dog, pig and cow based on sequence homology.

UNC5B Polyclonal antibody specifically detects UNC5B in Human samples. It is validated for Western Blot.
Specifications

Specifications

UNC5B
Polyclonal
Unconjugated
UNC5B
6330415E02Rik; A630020F16; C. elegans; D10Bwg0792e; Netrin receptor UNC5B; p53RDL1; p53-regulated receptor for death and life protein 1; Protein unc-5 homolog 2; protein unc-5 homolog B; transmembrane receptor Unc5H2; unc5 homolog 2; unc-5 homolog 2; unc-5 homolog B; unc-5 homolog B (C. elegans); unc-5 netrin receptor B; Unc5b; UNC5H2; UNQ1883/PRO4326
Goat
Antigen affinity chromatography
RUO
219699
-20°C
Liquid
Western Blot
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Q8IZJ1
UNC5B
Peptide with sequence C-SARDWIFQLKTQAHQ, from the internal region of UNC5B (aa629-643)
100 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions