missing translation for 'onlineSavingsMsg'
Learn More

ANTU, TRC

Product Code. 30335511
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30335511 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30335511 Supplier Toronto Research Chemicals Supplier No. TRCA6372231G
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Antu
CAS 86-88-4
Molecular Formula C11 H10 N2 S
Quantity 1 g
Synonym Thiourea, 1-naphthalenyl- (9CI), Urea, 1-(1-naphthyl)-2-thio- (7CI,8CI), N-1-Naphthalenylthiourea, 1-(1-Naphthyl)-2-thiourea, 1-(1-Naphthyl)thiourea, 1-Naphthylthiourea, ANTU, Alrato, Anturat, Bantu, Chemical 109, Dirax, Kripid, Krysid, N-1-Naphthylthiourea, NSC 3287, Naphthalen-1-ylthiourea, Naphtox, Rattrack, Smeesana, U 5227, α-Naphthylthiocarbamide, α-Naphthylthiourea
InChI Formula InChI=1S/C11H10N2S/c12-11(14)13-10-7-3-5-8-4-1-2-6-9(8)10/h1-7H,(H3,12,13,14)
SMILES NC(=S)Nc1cccc2ccccc12
IUPAC Name naphthalen-1-ylthiourea
Molecular Weight (g/mol) 202.28
Formula Weight 202.0565
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.