missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANXA2R Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23838-1-AP-150UL
This item is not returnable.
View return policy
Description
C5orf39 may act as a receptor for annexin II on marrow stromal cells to induce osteoclast formation.Specifications
| ANXA2R | |
| Polyclonal | |
| Unconjugated | |
| ANXA2R | |
| Annexin 2 receptor, Annexin II receptor, AX2R, AXIIR, C5orf39 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 389289 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q3ZCQ2 | |
| ANXA2R | |
| ANXA2R Fusion Protein Ag20807 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction