missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AOF1 Antibody (5E10C1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
217.00€ - 578.00€
Specifications
| Antigen | AOF1 |
|---|---|
| Clone | 5E10C1 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18277090
|
Novus Biologicals
NBP2-52415 |
0.1 mg |
578.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18611279
|
Novus Biologicals
NBP2-52415-0.025mg |
0.025 mg |
217.00€
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AOF1 Monoclonal specifically detects AOF1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| AOF1 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human | |
| AOF1, bA204B7.3, C6orf193, dJ298J15.2, DKFZp686I0412, flavin containing 1, flavin-containing amine oxidase domain-containing protein 1, FLJ33898, FLJ34109, FLJ43328, LSD2, lysine (K)-specific demethylase 1B, lysine-specific histone demethylase 1B, lysine-specific histone demethylase 2 | |
| KDM1B | |
| IgG1 | |
| Protein A or G purified |
| 5E10C1 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS with 0.05% Sodium Azide | |
| 221656 | |
| Purified recombinant fragment of human AOF1 (AA: 6-129) expressed in E. Coli. | |
| Primary | |
| 92 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title