missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCTAIRE2 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-42688IR
This item is not returnable.
View return policy
Description
PCTAIRE2 Polyclonal antibody specifically detects PCTAIRE2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| PCTAIRE2 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| Cell division protein kinase 17, cyclin-dependent kinase 17, EC 2.7.11, EC 2.7.11.22, PCTAIRE protein kinase 2, PCTAIRE2, PCTAIRE-motif protein kinase 2, PCTK2, protein kinase cdc2-related PCTAIRE-2, Serine/threonine-protein kinase PCTAIRE-2 | |
| The immunogen for this product maps to a region between residue 1 and 50 of human PCTAIRE protein kinase 2 using the numbering given in entry NP_002586.2 (GeneID 5128). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 5128 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction