Learn More
AP1G2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17907-1-AP-20UL
This item is not returnable.
View return policy
Specifications
| AP1G2 | |
| Polyclonal | |
| Unconjugated | |
| AP1G2 | |
| AP1G2, G2AD, Gamma2 adaptin | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100362547, 11766, 8906 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O75843, O88512 | |
| AP1G2 | |
| AP1G2 Fusion Protein Ag12300 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |