missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AP2 alpha Antibody (SR1826), Novus Biologicals™
Rabbit Monoclonal Antibody
338.00€ - 449.00€
Specifications
| Antigen | AP2 alpha |
|---|---|
| Clone | SR1826 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18644444
|
Novus Biologicals
NBP3-22200-100ul |
100 μg |
449.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18651214
|
Novus Biologicals
NBP3-22200-50ul |
50 μg |
338.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AP2 alpha Monoclonal antibody specifically detects AP2 alpha in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, ImmunoprecipitationSpecifications
| AP2 alpha | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Activating enhancer-binding protein 2-alpha, Activator protein 2, AP-2 transcription factor, AP2-alpha, AP2TFAP-2alpha, AP-2transcription factor AP-2 alpha (activating enhancer-binding protein 2 alpha), BOFS, FLJ51761, TFAP2, transcription factor AP-2 alpha (activating enhancer binding protein 2 alpha), transcription factor AP-2-alpha | |
| A synthesized peptide derived from human AP2 alpha (Uniprot #: P05549) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1826 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Cancer, Transcription Factors and Regulators | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 7020 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title