missing translation for 'onlineSavingsMsg'
Learn More

Apalutamide, TRC

Product Code. 30137727
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137727 25 mg 25mg
30135367 100 mg 100mg
18004020 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137727 Supplier Toronto Research Chemicals Supplier No. TRCA72612025MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 956104-40-8
Molecular Formula C21 H15 F4 N5 O2 S
Molecular Weight (g/mol) 477.4347
Synonym Benzamide, 4-[7-[6-cyano-5-(trifluoromethyl)-3-pyridinyl]-8-oxo-6-thioxo-5,7-diazaspiro[3.4]oct-5-yl]-2-fluoro-N-methyl-, 4-[7-[6-Cyano-5-(trifluoromethyl)-3-pyridinyl]-8-oxo-6-thioxo-5,7-diazaspiro[3.4]oct-5-yl]-2-fluoro-N-methylbenzamide, 4-[7-[6-Cyano-5-(trifluoromethyl)pyridin-3-yl]-8-oxo-6-thioxo-5,7-diazaspiro[3.4]octan-5-yl]-2-fluoro-N-methylbenzamide, A 52, AR 509, ARN 509, Apalutamide, Benzamide, 4-[7-[6-cyano-5-(trifluoromethyl)-3-pyridinyl]-8-oxo-6-thioxo-5,7-diazaspiro[3.4]oct-5-yl]-2-fluoro-N-methyl-, 4-[7-[6-Cyano-5-(trifluoromethyl)-3-pyridinyl]-8-oxo-6-thioxo-5,7-diazaspiro[3.4]oct-5-yl]-2-fluoro-N-methylbenzamide, 4-[7-[6-Cyano-5-(trifluoromethyl)pyridin-3-yl]-8-oxo-6-thioxo-5,7-diazaspiro[3.4]octan-5-yl]-2-fluoro-N-methylbenzamide, A 52, AR 509, ARN 509
IUPAC Name 4-[7-[6-cyano-5-(trifluoromethyl)pyridin-3-yl]-8-oxo-6-sulfanylidene-5,7-diazaspiro[3.4]octan-5-yl]-2-fluoro-N-methylbenzamide
SMILES CNC(=O)c1ccc(cc1F)N2C(=S)N(C(=O)C23CCC3)c4cnc(C#N)c(c4)C(F)(F)F

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C21H15F4N5O2S/c1-27-17(31)13-4-3-11(8-15(13)22)30-19(33)29(18(32)20(30)5-2-6-20)12-7-14(21(23,24)25)16(9-26)28-10-12/h3-4,7-8,10H,2,5-6H2,1H3,(H,27,31)
Formula Weight 477.0883
Chemical Name or Material Apalutamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.