missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APC Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-91662SS
This item is not returnable.
View return policy
Description
APC Polyclonal specifically detects APC in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| APC | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| adenomatosis polyposis coli, adenomatosis polyposis coli tumor suppressor, adenomatous polyposis coli, adenomatous polyposis coli protein, BTPS2, Deleted in polyposis 2.5, DP2.5DP2, DP3, FPC, GS, Protein APC | |
| Rabbit | |
| 310 kDa | |
| 0.025 mg | |
| Cancer, Cell Biology, Cell Cycle and Replication, Tumor Suppressors, Wnt Signaling Pathway | |
| 324 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1:500-1:1000, Flow Cytometry 2-10 ug/million cells, Immunohistochemistry 1:50-1:500, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200 | |
| P25054 | |
| APC | |
| Synthetic peptide made to the human APC protein (between amino acids 2750-2843) [UniProt P25054]. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction