Learn More
R&D Systems™ APE1 Redox Inhibitor, E3330
Product Code. 18626528 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18626528 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18626528 Supplier R&D Systems™ Supplier No. NBP14958125mg
  • ONLINE EXCLUSIVE PRICE
    730.94€
    643.65€ / 25mg
    Save 87.29€
    11% Off
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
A Novus product, part of R&D Systems.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at -20°C in powder form. Store at -80°C once reconstituted.
Product Line Novus
Quantity 25 mg
Type Inhibitor