missing translation for 'onlineSavingsMsg'
Learn More

Apigenin-7-O-sulfate Potassium Salt, TRC

Product Code. 30137728
Change view
Click to view available options
Quantity:
1 mg
25 mg
Unit Size:
1mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137728 25 mg 25mg
30005067 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137728 Supplier Toronto Research Chemicals Supplier No. TRCA72652025MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120562-19-8
Molecular Formula C15H9KO8S
Molecular Weight (g/mol) 388.39
Synonym 5-Hydroxy-2-(4-hydroxyphenyl)-7-(sulfooxy)-4H-1-benzopyran-4-one Potassium Salt;
IUPAC Name potassium;[5-hydroxy-2-(4-hydroxyphenyl)-4-oxochromen-7-yl] sulfate
SMILES [K+].Oc1ccc(cc1)C2=CC(=O)c3c(O)cc(OS(=O)(=O)[O-])cc3O2

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C15H10O8S.K/c16-9-3-1-8(2-4-9)13-7-12(18)15-11(17)5-10(6-14(15)22-13)23-24(19,20)21;/h1-7,16-17H,(H,19,20,21);/q;+1/p-1
Formula Weight 387.9655
Chemical Name or Material Apigenin-7-O-sulfate Potassium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.