Learn More
APOC1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16775-1-AP-150UL
 View more versions of this product
Product Code. 16806514
  • 410.00€ / 150µL
Description
The APOC1 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human APOC1. This antibody recognizes human antigen. The APOC1 antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
Specifications
APOC1
Polyclonal
Unconjugated
150 μL
341
IgG
Immunohistochemistry, ELISA
333 μg/mL