Learn More
APOC1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16775-1-AP-150UL
Description
The APOC1 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human APOC1. This antibody recognizes human antigen. The APOC1 antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
| APOC1 | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 341 | |
| IgG |
| Immunohistochemistry, ELISA | |
| 333 μg/mL | |
| Rabbit | |
| Primary | |
| Human |