Learn More
APOL2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25925-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene is a member of the apolipoprotein L gene family. The encoded protein is found in the cytoplasm, where it may affect the movement of lipids or allow the binding of lipids to organelles. Two transcript variants encoding the same protein have been found for this gene.Specifications
| APOL2 | |
| Polyclonal | |
| Unconjugated | |
| APOL2 | |
| APOL II, APOL2, APOL3, Apolipoprotein L II, apolipoprotein L, 2, Apolipoprotein L2 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 23780 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.53 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9BQE5 | |
| APOL2 | |
| APOL2 Fusion Protein Ag7622 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |