Learn More
Apolipoprotein A-IV/ApoA4 Antibody (CL0465) [Alexa Fluor™ 405], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52930AF405
Product Code. 30299245
  • 737.00€ / 0.10mL
Description
Apolipoprotein A-IV/ApoA4 Monoclonal antibody specifically detects Apolipoprotein A-IV/ApoA4 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Apolipoprotein A-IV/ApoA4
Monoclonal
Alexa Fluor 405
apo-AIV, ApoA-IV, Apolipoprotein A4, apolipoprotein A-IV, MGC142154, MGC142156
This antibody was developed using a recombinant protein derived from P06727, with the exact immunogen sequence remaining proprietary.
0.1 mL
Lipid and Metabolism
337
Store at 4⪚C in the dark.
IgG2a
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL0465
50mM Sodium Borate
Mouse
Protein A purified