missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apolipoprotein L1 Antibody (CL0173) [DyLight 488], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52871G
This item is not returnable.
View return policy
Description
Apolipoprotein L1 Monoclonal antibody specifically detects Apolipoprotein L1 in Human samples. It is validated for Western Blot
Specifications
| Apolipoprotein L1 | |
| Monoclonal | |
| DyLight 488 | |
| ApoL, APO-L, ApoL-I, Apolipoprotein L, apolipoprotein L, 1, apolipoprotein L1, Apolipoprotein L-I, FSGS4 | |
| This antibody was developed using a recombinant protein derived from O14791, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer, Cardiovascular Biology, Endocrinology | |
| 8542 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Western Blot | |
| CL0173 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido