missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apoptosis enhancing nuclease Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-31700
This item is not returnable.
View return policy
Description
Apoptosis enhancing nuclease Polyclonal specifically detects Apoptosis enhancing nuclease in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Apoptosis enhancing nuclease | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| AEN | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human Apoptosis-enhancing nuclease. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 64782 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.9mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| apoptosis enhancing nuclease, EC 3.1, EC 3.1.13.1, FLJ12484, FLJ12562, interferon stimulated exonuclease gene 20kDa-like 1, Interferon-stimulated 20 kDa exonuclease-like 1, ISG20L1apoptosis-enhancing nuclease, pp12744 | |
| Rabbit | |
| 36 kDa | |
| 0.1 mL | |
| Primary | |
| Tested in human. Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction