missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APPBP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14863-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene binds to the beta-amyloid precursor protein. Beta-amyloid precursor protein is a cell surface protein with signal-transducing properties, and it is thought to play a role in the pathogenesis of Alzheimer′s disease. In addition, the encoded protein can form a heterodimer with UBE1C and bind and activate NEDD8, a ubiquitin-like protein. This protein is required for cell cycle progression through the S/M checkpoint. Three transcript variants encoding different isoforms have been found for this gene.Specifications
| APPBP1 | |
| Polyclonal | |
| Unconjugated | |
| NAE1 | |
| A 116A10.1, APP BP1, APPBP1, HPP1, NAE1, Proto oncogene protein 1, ula 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 234664, 84019, 8883 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.28 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q13564, Q8VBW6, Q9Z1A5 | |
| NAE1 | |
| APPBP1 Fusion Protein Ag6672 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction