Learn More
Applied Biosystems™ MicroAmp™ 12-Cap Strip
Descripción
MicroAmp™ Cap Strips are designed to fit on MicroAmp™ reaction tubes, tube strips, and 96-well plates. They are available in 8-cap and 12-cap strips in clear or an assorted set of red, orange, blue, and green. They have a domed profile and are suitable for end-point PCR. Attach caps using the MicroAmp™ Cap Installing Tool.
Will not interfere with sample reading.
- Provides tight seal to minimize evaporation
- Easy to apply and remove
Minimizes Evaporation
Use MicroAmp™ Caps Strips to seal the tubes containing your precious samples. This will reduce the possibility of contamination among sample wells and help ensure reliable results.
Easy to Use
Use the MicroAmp™ Cap Installing Tool to push the caps down and form a tight seal. When ready, just peel away the caps using the tabs at the end.
Colored for Easy Organization
MicroAmp™ Caps Strips are available in different colors to help you keep track of your tubes during assay preparation.
Note: See user's manual or package insert for limited label license and trademark information.
Especificaciones
Especificaciones
| Certifications/Compliance | Certified DNA/Rnase Free |
| Color | Clear |
| Quantity | 200 Strips |
| For Use With (Equipment) | Thermal Cyclers |
| Product Line | MicroAmp™ |
| Material | Polypropylene |
| Content And Storage | Includes 200 MicroAmp™ 12-Cap Strips, Clear. Store at room temperature. |
| Reaction Speed | Standard |
| Shipping Condition | Room Temperature |
| Closure Type | Domed Cap |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.