missing translation for 'onlineSavingsMsg'
Learn More

Apraclonidine Hydrochloride, TRC

Product Code. 30179948
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179948 25 mg 25mg
30180354 10 mg 10mg
30180358 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179948 Supplier Toronto Research Chemicals Supplier No. TRCA72940025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73218-79-8
Molecular Formula C9 H10 Cl2 N4 . Cl H
Molecular Weight (g/mol) 281.57
Synonym 1,4-Benzenediamine, 2,6-dichloro-N1-(4,5-dihydro-1H-imidazol-2-yl)-, hydrochloride (1:1), 1,4-Benzenediamine, 2,6-dichloro-N-(4,5-dihydro-1H-imidazol-2-yl)-, monohydrochloride (9CI), 1,4-Benzenediamine, 2,6-dichloro-N1-(4,5-dihydro-1H-imidazol-2-yl)-, monohydrochloride, AL 02145, ALO 2145, Aplonidine hydrochloride, Apraclonidine hydrochloride, Iopidine, p-Aminoclonidine monohydrochloride
IUPAC Name 2,6-dichloro-1-N-(4,5-dihydro-1H-imidazol-2-yl)benzene-1,4-diamine;hydrochloride
SMILES Cl.Nc1cc(Cl)c(NC2=NCCN2)c(Cl)c1

Specifications

Analyte or Component Names Apraclonidine Hydrochloride
Shipping Condition Room Temperature
InChI Formula InChI=1S/C9H10Cl2N4.ClH/c10-6-3-5(12)4-7(11)8(6)15-9-13-1-2-14-9;/h3-4H,1-2,12H2,(H2,13,14,15);1H
Solution Type Neat
Industry Type Pharmaceutical
Quantity 25 mg
Formula Weight 280.0049
Chemical Name or Material Apraclonidine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.