missing translation for 'onlineSavingsMsg'
Learn More

Aprindine Hydrochloride, TRC

Product Code. 17920925
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17920925 250 mg 250mg
17944715 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17920925 Supplier Toronto Research Chemicals Supplier No. TRCA729900250MG
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33237-74-0
Molecular Formula C22 H30 N2 . Cl H
Molecular Weight (g/mol) 358.95
Synonym 1,3-Propanediamine, N1-(2,3-dihydro-1H-inden-2-yl)-N3,N3-diethyl-N1-phenyl-, hydrochloride (1:1), 1,3-Propanediamine, N,N-diethyl-N'-2-indanyl-N'-phenyl-, monohydrochloride (8CI), 1,3-Propanediamine, N-(2,3-dihydro-1H-inden-2-yl)-N',N'-diethyl-N-phenyl-, monohydrochloride (9CI), Amidonal, Aprindine hydrochloride, Aspenon, Compd 83846, Fibocil, Fiboran, MS 5075, NSC 284614, Ritmusin
IUPAC Name N'-(2,3-dihydro-1H-inden-2-yl)-N,N-diethyl-N'-phenylpropane-1,3-diamine;hydrochloride
SMILES Cl.CCN(CC)CCCN(C1Cc2ccccc2C1)c3ccccc3

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C22H30N2.ClH/c1-3-23(4-2)15-10-16-24(21-13-6-5-7-14-21)22-17-19-11-8-9-12-20(19)18-22;/h5-9,11-14,22H,3-4,10,15-18H2,1-2H3;1H
Formula Weight 358.2176
Chemical Name or Material Aprindine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.