Learn More
Veolia Water Systems™ Aquadem™ Regeneration Bottle
Product Code. 11913266
Change view
Click to view available options
Capacity (Metric):
8 L
50 L
60 L
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11913266 8 L Each
11933266 50 L Each
11943266 60 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11913266 Supplier Veolia Water Systems™ Supplier No. AARIXD200694
  • ONLINE EXCLUSIVE PRICE
    290.85€
    227.14€ / Pack of 1
    Save 63.71€
    21% Off
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Rich, robust, regeneration bottle for pre-treatment and polishing systems.
  • AQUADEM regeneration stations and filling centers ensure the treatment of resins
  • Each batch is subjected to a strict quality control, with recording of the regeneration
Specifications
Capacity (Metric) 8 L